![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ALB |
Antigen Name | Human Serum Albumin |
Reactivity | Mouse, Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
Antigen Synonyms | ANALBA, Cell Growth Inhibiting Protein 42, serum albumin, FDAH, PRO0883, PRO1341, PRO0903, Growth-Inhibiting Protein 20 |
Gene ID | 213 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |