Anti-ALAS2 Mouse Monoclonal Antibody [clone: 6C1]

Supplier: Abnova
H00000212-M01
10576-632EA 335 USD
10576-632
Anti-ALAS2 Mouse Monoclonal Antibody [clone: 6C1]
Antibodies
Type: Primary
Antigen: ALAS2
Clonality: Monoclonal
Clone: 6C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR