Anti-AKT2 Mouse Monoclonal Antibody [clone: X1]

Supplier: Abnova
H00000208-M06
10576-608EA 335 USD
10576-608
Anti-AKT2 Mouse Monoclonal Antibody [clone: X1]
Antibodies
Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: X1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR