Anti-AKT2 Mouse Monoclonal Antibody [clone: 1D9]

Supplier: Abnova
H00000208-M03
10576-604EA 335 USD
10576-604
Anti-AKT2 Mouse Monoclonal Antibody [clone: 1D9]
Antibodies
Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR