Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Mouse Monoclonal Antibody [clone: 3E11]

Supplier: Abnova
H00000207-M26
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 3E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg H00000207-M26 10576-580 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol AKT1
Antigen Name V-Akt Murine Thymoma Viral Oncogene Homolog 1
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 3E11
Host Mouse
Western Blot Yes
ELISA Yes
Antigen Synonyms C-AKT, rac protein kinase beta, RAC, RAC-gamma serine/threonine protein kinase, EC 2.7.11.1, RAC-alpha serine/threonine kinase, PKB, RAC-gamma, Protein kinase B beta, RAC-beta serine/threonine-protein kinase, RAC-PK-alpha, Protein kinase Akt-2, PKB-alpha, Murine thymoma viral (v-akt) homolog-2, PKB-GAMMA, PRKBG, AKT, PKBBETA, PKB beta, AKT1 kinase, kinase Akt1, RAC-BETA, v-akt murine thymoma viral oncogene homolo, RAC-PK-beta, STK-2, PKB gamma, PRKBB, PKBB, RAC-gamma serine/threonine-protein kinase, PKBG, Protein kinase B, RAC-PK-gamma, AKT2
Gene ID 207
Cross Adsorption No
Storage Temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty