Anti-AKT1 Mouse Monoclonal Antibody [clone: 2E11]

Supplier: Abnova
H00000207-M03
10576-584EA 335 USD
10576-584
Anti-AKT1 Mouse Monoclonal Antibody [clone: 2E11]
Antibodies
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR