Anti-AK1 Mouse Monoclonal Antibody [clone: 4C2-1A8]

Supplier: Abnova
H00000203-M07
10576-484EA 335 USD
10576-484
Anti-AK1 Mouse Monoclonal Antibody [clone: 4C2-1A8]
Antibodies
Type: Primary
Antigen: AK1
Clonality: Monoclonal
Clone: 4C2-1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR