Anti-AHR Mouse Monoclonal Antibody [clone: 3B7]

Supplier: Abnova
H00000196-M09
10576-454EA 335 USD
10576-454
Anti-AHR Mouse Monoclonal Antibody [clone: 3B7]
Antibodies
Type: Primary
Antigen: AHR
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR