Anti-AHR Mouse Monoclonal Antibody [clone: 4G7]

Supplier: Abnova
H00000196-M04
10576-458EA 335 USD
10576-458
Anti-AHR Mouse Monoclonal Antibody [clone: 4G7]
Antibodies
Type: Primary
Antigen: AHR
Clonality: Monoclonal
Clone: 4G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR