Anti-AHCY Mouse Monoclonal Antibody [clone: M2]

Supplier: Abnova
H00000191-M09
10576-428EA 335 USD
10576-428
Anti-AHCY Mouse Monoclonal Antibody [clone: M2]
Antibodies
Type: Primary
Antigen: AHCY
Clonality: Monoclonal
Clone: M2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR