Anti-NR0B1 Mouse Monoclonal Antibody [clone: 3G8]

Supplier: Abnova
H00000190-M07
10595-292EA 335 USD
10595-292
Anti-NR0B1 Mouse Monoclonal Antibody [clone: 3G8]
Antibodies
Type: Primary
Antigen: NR0B1
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR