Anti-AGER Mouse Monoclonal Antibody [clone: 1C1]

Supplier: Abnova
H00000177-M02C
10612-338EA 335 USD
10612-338
Anti-AGER Mouse Monoclonal Antibody [clone: 1C1]
Antibodies
Type: Primary
Antigen: AGER
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR