Anti-UBR5 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2101B
80088-422EA 751.67 USD
80088-422
Anti-UBR5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UBR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR