Anti-SNAI1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2054B
60941-458EA 789.25 USD
60941-458
Anti-SNAI1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SNAI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal R8
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR