Anti-SNAI2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2053A
80088-396EA 789.68 USD
80088-396
Anti-SNAI2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SNAI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR