Anti-ZFP42 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2051A
80088-390EA 789.68 USD
80088-390
Anti-ZFP42 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZFP42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–50 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR