Anti-NEUROG1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2022A
80088-338EA 789.25 USD
80088-338
Anti-NEUROG1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NEUROG1 (neurogenin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR