Anti-PDPN Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2016B
80088-316EA 789.25 USD
80088-316
Anti-PDPN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 48–63 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR