Anti-NUP50 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1913A
60941-404EA 751.67 USD
60941-404
Anti-NUP50 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NUP50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR