Anti-GAS41 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1910A
60941-402EA 789.25 USD
60941-402
Anti-GAS41 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GAS41 (YEATS4/NuBI-1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR