Anti-UPF1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1905C
60941-384EA 789.68 USD
60941-384
Anti-UPF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UPF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR