Anti-AGO2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1903A
60941-376EA 789.25 USD
60941-376
Anti-AGO2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AGO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR