Anti-DICER1 (dicer 1, ribonuclease type III) Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1901C
60941-372EA 751.67 USD
60941-372
Anti-DICER1 (dicer 1, ribonuclease type III) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DICER1 (dicer 1, ribonuclease type III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR