Anti-RAB24 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1819A
60941-350EA 751.67 USD
60941-350
Anti-RAB24 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAB24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR