Anti-ATG16L1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1817C
60941-342EA 796.63 USD
60941-342
Anti-ATG16L1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATG16L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR