Anti-ATG9A Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1814B
60941-324EA 789.25 USD
60941-324
Anti-ATG9A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATG9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR