Anti-ATG7 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1813A
60941-314EA 789.68 USD
60941-314
Anti-ATG7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATG7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR