Anti-ATG5 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1812B
60941-312EA 791.93 USD
60941-312
Anti-ATG5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR