Anti-ATG4C Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1810C
60941-302EA 785.02 USD
60941-302
Anti-ATG4C Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATG4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR