Anti-ATG3 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1807B
60941-282EA 789.25 USD
60941-282
Anti-ATG3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR