Anti-LC3B Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1806A
60941-278EA 789.68 USD
60941-278
Anti-LC3B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR