Anti-LC3 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1802B
60941-268EA 789.68 USD
60941-268
Anti-LC3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR