Anti-BMP6 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1717A
80088-256EA 789.68 USD
80088-256
Anti-BMP6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BMP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–60 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR