Anti-IL28 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1708B
80088-236EA 789.25 USD
80088-236
Anti-IL28 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center ( 1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR