Anti-SIGLEC7 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1633D
80088-222EA 751.67 USD
80088-222
Anti-SIGLEC7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SIGLEC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal G391
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR