Anti-GFP Tag Mouse Monoclonal Antibody

Supplier: Abcepta
AM1009A
80087-654EA 813.39 USD
80087-654
Anti-GFP Tag Mouse Monoclonal Antibody
Antibodies
Type: Tag
Antigen: GFP Tag
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR