Would you like to log in again?
WebSeal™ 384-Well Non-Coated Plastic Shallow Well Microplates, Thermo Scientific
Supplier: Thermo Scientific
60180-P136 60180-P106
10036-132CS 910.47 USD InStock
10036-132 10036-130
WebSeal™ 384-Well Non-Coated Plastic Shallow Well Microplates, Thermo Scientific
Microplates Solid Supported Liquid/Liquid Extraction Plates (SLE)
WebSeal non-coated, plastic microplates have excellent solvent resistance and low background noise
Made of high-quality, GC-tested polypropylene. Ideal for pharmaceutical and industrial applications, as well as high-throughput screening, sample collection and storage, combinatorial chemistry and liquid phase assays.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty