Trap Column Cartridge Holder with nanoViper Fittings, Thermo Scientific
Supplier: Thermo Scientific
164649
10067-728EA 589.26 USD
10067-728
Trap Column Cartridge Holder with nanoViper Fittings, Thermo Scientific
Racks Chromatography Vial Racks
μ-Precolumn holder, 5 mm, with 30 μm I.D connecting tubing, nanoViper fittings.
For trap cartridges with 2 x 100 mm length nanoViper fittings.