Sodium phosphate dibasic, anhydrous USP, FCC, Ph. Eur.

Synonyms: di-Sodium hydrogen phosphatedi-Sodium hydrogen orthophosphate

51115000120 51115000500
10755-340EA 2104.77 USD
10755-340 10755-746
Sodium phosphate dibasic, anhydrous USP, FCC, Ph. Eur.
di-Sodium hydrogen phosphate

For excipient use only.


Certifications: Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.


Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.

Formula: Na₂HPO₄
MW: 141.96 g/mol
MDL Number: MFCD00003496
CAS Number: 7558-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR