Alere iScreen® DX Dip Card, CLIA-Waived Multi-Panel Drug Test Kits, Alere™ Toxicology
IS10-DX IS12-DX IS10-22015-DX IS10BUP-DX IS5 M-DX
89423-360PK 321.74 USD
89423-360 89423-362 10753-242 89423-358 89423-364
Alere iScreen® DX Dip Card, CLIA-Waived Multi-Panel Drug Test Kits, Alere™ Toxicology
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Dip Card Format. Designed to screen multiple drug metabolites in one simple dip and read procedure with results in 5 minutes.
Room temperature storage, long shelf-life. Collection cup not included.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI) and (MOP), Methadone (MTD), Oxycodone (OXY), Phencyclidine (PCP), Propoxyphene (PPX), Ecstacy (MDMA), Tricyclic Antidepressents (TCA), and Marijuana (THC) in human urine at the following cutoff levels (ng/mL) unless otherwise noted in description: AMP (1000), BAR (300), BZO (300), BUP (10), COC (300), mAMP (1000), OPI (2000), MOP (300), MTD (300), OXY (100), PCP (25), PPX (300), MDMA (500), TCA (1000), and THC (50).
Ordering information: The iScreen® Control Kit (item #88004) can be used with the following iScreen® Dx Multi-Panel dip cards. Kit includes two vials (5mL positive and 5mL negative external controls). Refrigerated, must be shipped next day (sold separately).