Cabinet with Two Removable Adjustable Trays, Vented Storage, Refrigerator, TrippNT
Supplier: TrippNT
50385
10012-846EA 1112.24 USD
10012-846
Cabinet with Two Removable Adjustable Trays, Vented Storage, Refrigerator, TrippNT
Racks Bottle Racks
  • Refrigerator or freezer storage with removable trays
  • Adjustable dividers create many compartments
  • Tray comes with 2 removable trays but holds 4
  • White PVC and acrylic construction
  • Comes with 2 dividers per tray, but each tray can hold 10 dividers
  • In stock and ready to ship
  • American made by TrippNT
  • Total replacement Lifetime Guarantee
Vented cabinet allows airflow for efficient cooling in a refrigerator or freezer. Comes with 2 trays with adjustable dividers. Cabinet will hold 4 trays however. Extra trays can be purchased separately. This 16 inch wide cabinet is 9 inches tall and has trays with adjustable dividers for flexible storage of small bottles, samples, boxed items or extra on the counter drawer space. Rust proof PVC and acrylic construction. Durable and attractive storage for large quantities of small bottles. White/Blue color.