Anti-LI Cadherin Rabbit Polyclonal Antibody

Supplier: Boster
PA2200
10209-302EA 809.04 USD
10209-302
Anti-LI Cadherin Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Cadherin-17(CDH17) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: LI Cadherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR