Anti-Caspase-8(P10) Rabbit Polyclonal Antibody

Supplier: Boster
PA1524
10209-166EA 809.04 USD
10209-166
Anti-Caspase-8(P10) Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Caspase-8(CASP8) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Caspase-8(P10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR