Anti-STIM1 Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J49
CA10037-266
Anti-STIM1 Polyclonal Antibody
Antibodies
Type: Primary
Antigen: STIM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR