Anti-RFP Chicken Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-906-379 600-906-379
RL600906379EA 644.25 USD
CA11029-172 RL600906379
Anti-RFP Chicken Polyclonal Antibody (Biotin)
Antibodies
Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR