Anti-HDAC11 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J17 600-401-J17
RL600401J17EA 518.16 USD
CA11029-136 RL600401J17
Anti-HDAC11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HDAC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR