Anti-Abeta PYRO E11 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J13 600-401-J13
RL600401J13EA 518.16 USD
RL600401J13 CA11029-170
Anti-Abeta PYRO E11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Abeta PYRO E11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR