Anti-IL22 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
209-406-J33 209-406-J33
RL209406J33EA 483.1 USD
RL209406J33 CA11029-168
Anti-IL22 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: IL22
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR