Anti-TSLP Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
209-406-H48 209-406-H48
RL209406H48EA 497.78 USD
RL209406H48 CA11029-164
Anti-TSLP Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: TSLP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR