Anti-RNASEA Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4688-0100 200-4688-0100
RL20046880100EA 497.78 USD
RL20046880100 CA11029-338
Anti-RNASEA Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: RNASEA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR